Learn More
YM-58483, MedChemExpress
Product Code. 16400046
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16400046 2 mg 2mg
16482656 25 mg 25mg
16454466 100 mg 100mg
16445917 5 mg 5mg
16439736 50 mg 50mg
16423188 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16400046 Supplier Medchem Express Supplier No. HY100831
MedChemExpress YM-58483 (BTP2) is the first selective and potent inhibitor of CRAC channels and subsequent Ca2+ signals. YM-584832 is a blocker of store-operated Ca2+ entry (SOCE).
YM-58483 (BTP2) is the first selective and potent inhibitor of CRAC channels and subsequent Ca2+ signals. YM-584832 is a blocker of store-operated Ca2+ entry (SOCE).
Specifications
Chemical Name or Material YM-58483
Physical Form Solid
CAS 223499-30-7
Color White
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C15H9F6N5OS
For Use With (Application) COVID-19-immunoregulation
Synonym BTP2
Solubility Information DMSO : 125 mg/mL (296.69 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N=NS1)NC2=CC=C(N3N=C(C(F)(F)F)C=C3C(F)(F)F)C=C2
Molecular Weight (g/mol) 421.32
Formula Weight 421.32
Percent Purity 98.75%
Grade Research