Learn More
Checkers Yellow Jacket™ 5 Channel Heavy Duty Cable Protector
Product Code. 17742428
Change view
Click to view available options
Color:
Black, Yellow
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17742428 Black, Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17742428 Supplier Checkers Supplier No. PYJS0001YB
  • ONLINE EXCLUSIVE PRICE
    427.17 EUR
    346.88 EUR / Pack of 1
    Save 80.29 EUR
    18% Off
Estimated Shipment: 03-12-2026
Yellow Jacket™ 5-channel cable protector with hinged lid provides a safer crossing for vehicles and pedestrian traffic while protecting valuable electrical cables and hose lines from damage.
  • Yellow Jacket™ 5-channel cable protector with hinged lid provides a safer crossing for vehicles and pedestrian traffic while protecting valuable electrical cables and hose lines from damage
Specifications
Length (English) 36 in.
Length (Metric) 91 cm
Width (Metric) 51 cm
Color Black, Yellow
Material Polyethylene
Type Cable Protector