missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Tetrasodium Salt, ACS Grade, Scharlab

Product Code. 11322645
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322645 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11322645 Supplier Scharlab Supplier No. AN00900005
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used in analytical chemistry and suitable as an indicator for metal titration.

  • Synonyms: 3',3-Bis[bis(carboxymethyl)-aminoethyl] cresol sulfone phtrane sodium salt
  • Molecular Formula: C31H28N2Na4O13S
  • Molar Mass: 760.60g/mol
  • CAS: 3618-43-7

Specifications

CAS 3618-43-7
Color Orange
Loss on Drying 7% max (110°C)
Molecular Formula C31H28N2Na4O13S
Quantity 5 g
Synonym 3',3'-Bis[bis(carboxymethyl)-aminoethyl] cresol sulfone phtrane sodium salt
Formula Weight 760.60g/mol
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.