missing translation for 'onlineSavingsMsg'
Learn More

Xylene, TRC

Product Code. 17972635
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972635 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 17972635 Supplier Toronto Research Chemicals Supplier No. TRCX749400100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1330-20-7
Quantity 100 g
Molecular Formula C8H10
Synonym Xylenes, Benzene, dimethyl-, Xylene (8CI), Dimethylbenzene, Dilan, Entellan New, Xylol, ZEP-RD, Methyltoluene, UN 1307
InChI Formula InChI=1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3
SMILES CO
IUPAC Name m-xylene--o-xylene--p-xylene (1/1/1)
Molecular Weight (g/mol) 106.17
Formula Weight 318.2348
Chemical Name or Material Xylene (all isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.