Learn More
Xylazine Hydrochloride, TRC
Product Code. 30010641
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010641 100 mg 100mg
30007524 1 g 1g
30009277 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010641 Supplier Toronto Research Chemicals Supplier No. TRCX748000100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23076-35-9
Molecular Formula C12 H16 N2 S . Cl H
Molecular Weight (g/mol) 256.79
Synonym Xylazine Hydrochloride, N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine monohydrochloride, 5,6-Dihydro-2-(2,6-xylidino)-4H-1,3-thiazine hydrochloride, BAY 1470 hydrochloride, BAY-Va 1470, Celactal, Narcosyl, Rompun, Xylapan, Xylasol, Xylazine chloride, Xylazine hydrochloride
IUPAC Name N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine;hydrochloride
SMILES Cl.Cc1cccc(C)c1NC2=NCCCS2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C12H16N2S.ClH/c1-9-5-3-6-10(2)11(9)14-12-13-7-4-8-15-12;/h3,5-6H,4,7-8H2,1-2H3,(H,13,14);1H
Formula Weight 256.0801
Chemical Name or Material Xylazine hydrochloride