missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPNPEP3 Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
Beskrivelse
XPNPEP3 Monoclonal antibody specifically detects XPNPEP3 in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Tekniske data
Tekniske data
| Antigen | XPNPEP3 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2D5 |
| Concentration | 0.81 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | ATG3 |
| Gene Accession No. | Q6AZ50, Q9CPX6, Q9NT62 |
| Gene Alias | Aminopeptidase P3; APP3; E430012M05Rik; ICP55; Intermediate Cleaving Peptidase 55; LOW QUALITY PROTEIN: xaa-Pro aminopeptidase 3; NPHPL1; probable Xaa-Pro aminopeptidase 3; putative Xaa-Pro aminopeptidase 3; Xaa-Pro aminopeptidase 3; xaa-Pro aminopeptidase 3; probable Xaa-Pro aminopeptidase 3; XPNPEP3; X-Pro aminopeptidase 3; X-prolyl aminopeptidase (aminopeptidase P) 3, putative; X-prolyl aminopeptidase 3; X-prolyl aminopeptidase 3, mitochondrial; zgc:76905; zgc:76905 protein |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?