missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ XMU MP 1
GREENER_CHOICE

Product Code. 18798066
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18798066

Brand: Tocris Bioscience™ 6482/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective ATP-competitive mammalian sterile 20-like kinase (MST) 1/2 inhibitor

XMU MP 1 is a potent and selective ATP-competitive mammalian sterile 20-like kinase (MST) 1/2 inhibitor (IC50 values are 38.1 and 71.1 nM for MST2 and MST1, respectively). Enhances activity of downstream YAP in vitro. Augments mouse intestinal repair and promotes liver repair and regeneration in a mouse model of acute and chronic liver injury. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[(6,10-Dihydro-5,10-dimethyl-6-oxo-5H-pyrimido[5,4-b]thieno[3,2-e][1,4]diazepin-2-yl)amino]benzenesulfonamide
CAS 2061980-01-4
Quantity 50 mg
Molecular Formula C17H16N6O3S2
Formula Weight 416.48
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.