missing translation for 'onlineSavingsMsg'
Learn More

XL041, MedChemExpress

Product Code. p-200101853
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470959 1 mg 1mg
16464746 100 mg 100mg
16460057 50 mg 50mg
16443668 10 mg 10mg
16416407 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16470959 Supplier Medchem Express Supplier No. HY101973
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress XL041 (BMS-852927) is an LXRβ-selective agonist.

XL041 (BMS-852927) is an LXRβ-selective agonist.

Specifications

Chemical Name or Material XL041
CAS 1256918-39-4
Color White
Purity Grade Notes Research
Molecular Formula C29H28Cl2F2N2O4S
Quantity 1 mg
Synonym BMS-852927
Solubility Information DMSO : 100 mg/mL (164.07 mM; Need ultrasonic)
SMILES OC(C)(C)C1=CN(C2=CC=C(C3=CC(S(=O)(C)=O)=C(CO)C(F)=C3)C=C2F)C(C(C)(C4=C(Cl)C=CC=C4Cl)C)=N1
Molecular Weight (g/mol) 609.51
Formula Weight 609.51
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.