Learn More
Xipamide, TRC
Specifications
Specifications
| Chemical Name or Material | Xipamide |
| CAS | 14293-44-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H15 Cl N2 O4 S |
| Quantity | 100 mg |
| Synonym | Benzamide','5-(aminosulfonyl)-4-chloro-N-(2,6-dimethylphenyl)-2-hydroxy-, 2',6'-Salicyloxylidide','4-chloro-5-sulfamoyl- (8CI), Aquaphor, Aquaphor (diuretic), BE 1293, BEI 1293, Chronexan, Diurex, Diurex (Lacer), Diurexan, Lumitens, MJF 10938, Xipamide |
| InChI Formula | InChI=1S/C15H15ClN2O4S/c1-8-4-3-5-9(2)14(8)18-15(20)10-6-13(23(17,21)22)11(16)7-12(10)19/h3-7,19H,1-2H3,(H,18,20)(H2,17,21,22) |
| SMILES | Cc1cccc(C)c1NC(=O)c2cc(c(Cl)cc2O)S(=O)(=O)N |
| IUPAC Name | 4-chloro-N-(2,6-dimethylphenyl)-2-hydroxy-5-sulfamoylbenzamide |
| Molecular Weight (g/mol) | 354.81 |
| Show More |