Learn More
Xipamide-d6, TRC
Product Code. 30455013
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455013 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30455013 Supplier Toronto Research Chemicals Supplier No. TRCX74515210MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Xipamide-d6
CAS 1330262-09-3
Molecular Formula C15H9D6ClN2O4S
Quantity 10 mg
Synonym 5-(Aminosulfonyl)-4-chloro-N-(2,6-dimethylphenyl)-2-hydroxy-benzamide-d6, 4-Chloro-5-sulfamoyl-2',6'-salicyloxylidide-d6, Aquaphor-d6, Chronexan-d6, Diurex-d6, Lumitens-d6
InChI Formula InChI=1S/C15H15ClN2O4S/c1-8-4-3-5-9(2)14(8)18-15(20)10-6-13(23(17,21)22)11(16)7-12(10)19/h3-7,19H,1-2H3,(H,18,20)(H2,17,21,22)/i1D3,2D3
SMILES [2H]C([2H])([2H])c1cccc(c1NC(=O)c2cc(c(Cl)cc2O)S(=O)(=O)N)C([2H])([2H])[2H]
Molecular Weight (g/mol) 360.85
Formula Weight 360.0818