missing translation for 'onlineSavingsMsg'
Learn More

Xenon Difluoride, Sigma-Aldrich™

Product Code. 15830671
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15830671 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15830671 Supplier Sigma Aldrich Supplier No. 8188880001
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 130°C to 135°C (Sublimed)
CAS 13709-36-9
Quantity 1 g
MDL Number MFCD00040538
Synonym Xenon difluoride, Xenon difluoride
Molecular Weight (g/mol) 169.29
Grade Research
Chemical Name or Material Xenon Difluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.