Learn More
Lovibond™ XD7500 UV-visible spectrophotometer
Product Code. 15954442
Change view
Click to view available options
For Use With (Application):
Spectrophotometry
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
15954442 Spectrophotometry Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15954442 Supplier Lovibond™ Supplier No. 71307500
Add to basket
Add to basket
ideal for routine and spectral analysis
The XD 7500 is state-of-the-art spectrophotometers. Due to many technical subtleties, such as The automatic test recognition via barcode or over 150 pre-programmed Lovibond™ methods makes it ideal for routine or spectral analysis.
Latest technology
The XD 7000 and XD 7500 spectrophotometers are equipped with VIS respectively UV / VIS reference beam technology.
Automatic test recognition
Each of the more than 150 pre-programmed Lovibond™ methods is recognized by barcode as well as the cuvette type.
Analytical quality assurance
Standard procedures for analytical quality assurance support the verification of the photometer, the overall system (including the chemical methodology) and the verification of matrix effects.
Built-in security levels
The devices support the assignment of passwords and the assignment of up to three different levels of rights.
Additional functions
Measurement of transmission and absorption, spectral scan, kinetics analysis, as well as the creation of user-defined methods.
Interfaces for data processing
You want to process your data? With Ethernet, USB B, USB A for external memory, keyboard, barcode scanner and printer different versions are available.
Specifications
Display 7 in. High Contrast Color Graphic
Light Source Xenon Flash
Spectral Bandwidth 190 to 1100 nm
For Use With (Application) Spectrophotometry
Optical Design Grid monochromator with reference beam and beam splitter after exit slit
Pathlength 10 to 50 mm
Photometric Range -3.3 A to +3.3 A
Wavelength Accuracy ±1 nm