Learn More
Trajan Scientific and Medical™ XCHANGE™ Syringes for use with MEPS BINS
Product Code. 15219273
Change view
Click to view available options
Volume (Metric):
100 μL
250 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15219273 250 μL Each
15289263 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15219273 Supplier Trajan Scientific and Medical™ Supplier No. 2928600
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
With PTFE-tipped plunger
  • High quality solutions for liquid handling
  • For use on CTC PAL-xt
Accurate and reproducible injectionsXCHANGE Syringes specifically designed for MEPS Applications
Specifications
Material PTFE (Plunger)
Diameter (Metric) Outer 6.7 mm syringe barrel
Volume (Metric) 250 μL