Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-xt Autosampler Syringes
Product Code. 15279263
Change view
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
Diameter (Metric) Outer Needle:
0.63 mm
0.71 mm
Material:
Metal (Plunger)
PTFE (Plunger)
PTFE-tipped (Plunger)
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Diameter (Metric) Outer Needle Material Unit Size
15279263 50 μL 0.71 mm PTFE-tipped (Plunger) Each
15249263 25 μL 0.63 mm PTFE (Plunger) Each
15259263 25 μL 0.71 mm PTFE-tipped (Plunger) Each
15219263 10 μL 0.63 mm Metal (Plunger) Each
15229263 10 μL 0.71 mm Metal (Plunger) Each
15239263 10 μL 0.63 mm PTFE (Plunger) Each
15269263 50 μL 0.63 mm PTFE-tipped (Plunger) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 missing translation for 'options'
Product Code. 15279263 missing translation for 'mfr' Trajan Scientific and Medical™ missing translation for 'supplierNo' 2928430
Estimated Shipment: 20-01-2027
Allows adding of internal standards without cross contamination
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Available as all-metal or PTFE-tipped plunger
Specifications
Material PTFE-tipped (Plunger)
Description Autosampler Syringe
Needle Gauge 22 G
Diameter (Metric) Inner Needle 0.17 mm
Diameter (Metric) Outer Needle 0.71 mm
Volume (Metric) 50 μL
Needle Point Style LC