Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-xt Autosampler Syringes
Product Code. 15249263
Change view
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
Diameter (Metric) Outer Needle:
0.63 mm
0.71 mm
Material:
Metal (Plunger)
PTFE (Plunger)
PTFE-tipped (Plunger)
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Diameter (Metric) Outer Needle Material unitSize
15249263 25 μL 0.63 mm PTFE (Plunger) Each
15259263 25 μL 0.71 mm PTFE-tipped (Plunger) Each
15219263 10 μL 0.63 mm Metal (Plunger) Each
15229263 10 μL 0.71 mm Metal (Plunger) Each
15239263 10 μL 0.63 mm PTFE (Plunger) Each
15269263 50 μL 0.63 mm PTFE-tipped (Plunger) Each
15279263 50 μL 0.71 mm PTFE-tipped (Plunger) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15249263 Supplier Trajan Scientific and Medical™ Supplier No. 2928310
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Allows adding of internal standards without cross contamination
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Available as all-metal or PTFE-tipped plunger
Specifications
Material PTFE (Plunger)
Description Autosampler Syringe
Needle Gauge 23 G
Diameter (Metric) Inner Needle 0.15 mm
Diameter (Metric) Outer Needle 0.63 mm
Volume (Metric) 25 μL
Needle Point Style Cone