Learn More
Xanthene-9-carboxylic Acid 2-(Diethylamino)ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Xanthene-9-carboxylic Acid 2-(Diethylamino)ethyl Ester |
| CAS | 24539-72-8 |
| Molecular Formula | C20H23NO3 |
| Quantity | 500 mg |
| Synonym | 9H-Xanthene-9-carboxylic Acid 2-(Diethylamino)ethyl Ester, NSC 108248, SC 920 |
| InChI Formula | InChI=1S/C20H23NO3/c1-3-21(4-2)13-14-23-20(22)19-15-9-5-7-11-17(15)24-18-12-8-6-10-16(18)19/h5-12,19H,3-4,13-14H2,1-2H3 |
| SMILES | CCN(CC)CCOC(=O)C1c2ccccc2Oc3ccccc13 |
| IUPAC Name | 2-(diethylamino)ethyl 9H-xanthene-9-carboxylate |
| Molecular Weight (g/mol) | 325.4 |
| Formula Weight | 325.1678 |