Learn More
Epredia X1000 Round Coverslip dia. 22mm #1 (0.13-0.16mm) .
Brand:  Epredia CB00220RA120MNZ0
X1000 Round Coverslip dia. 22mm #1 (0.13-0.16mm) .
Product Code. 10015871
  • ONLINE EXCLUSIVE PRICE
    74.35 EUR
    66.06 EUR / Pack of 1000
    Save 8.29 EUR
    11% Off
 Restricted Item
Estimated Shipment: 12-10-2026