missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WZB 117
GREENER_CHOICE

Product Code. 18786606
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18786606

Brand: Tocris Bioscience™ 6143/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GLUT inhibitor

WZB 117 is a GLUT inhibitor (Ki(app) values are 0.2, 10 and 10 μM for GLUT4, GLUT1 and GLUT3, respectively). Competitive inhibitor of net glucose uptake but a noncompetitive inhibitor of sugar efflux from cells. Inhibits passive sugar transport in human erythrocytes and cancer cell lines. Attenuates erythrocyte 3-O-metylglucose (3MG) uptake in vitro (Ki = 6 μM). Suppresses tumor growth in a nude mouse model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Fluoro-1,2-phenylene bis(3-hydroxybenzoate)
CAS 1223397-11-2
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C20H13FO6
Formula Weight Observed MW: 368.31
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.