Learn More
Cole Parmer™ Write-On Tape
Product Code. 15366155 Shop All Cole Parmer Products
Change view
Click to view available options
Color:
Assorted
White
Width (English):
0.5 in.
Unit Size:
Pack of 24
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Width (English) Unit Size
15366155 Assorted 0.5 in. Pack of 24
15719404 White 0.5 in. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15366155 Supplier Cole Parmer™ Supplier No. 0653028
  • ONLINE EXCLUSIVE PRICE
    78.45 EUR
    76.32 EUR / Pack of 24
    Save 2.13 EUR
    2% Off
Estimated Shipment: 15-12-2026
Use to label/identify laboratory items/products.
  • Identify with writing and color.
  • Durable tape is waterproof, and oil and acid resistant.
  • Can be used with pencil, pen, or permanent marker.
  • Temperature resistant.
Specifications
Color Assorted
No. per Pack 24/Pk.
Quantity 24/Pk.
Temperature Range (Metric) -23°C to +121°C
Dimensions (L x W) 14 yd. x 0.5 in.
Length (English) 14 yd.
Width (English) 0.5 in.