Learn More
3M™ WP96 Series Faceshield, Polycarbonate, Clear, 10 ea/Case
Product Code. 15739067 Shop All 3M Products
Change view
Click to view available options
Size: 230 x 370mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15739067 Size: 230 x 370mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15739067 Supplier 3M™ Supplier No. 7100016704
  • ONLINE EXCLUSIVE PRICE
    157.00 EUR
    110.07 EUR / Pack of 10
    Save 46.93 EUR
    29% Off
Estimated Shipment: 11-01-2027
3M™ WP96 Series Faceshield is our high impact resistant, clear polycarbonate faceshield designed to fit 3M™ Hard Hats. It can be used with H8 Headgear easy faceshield replacement system.
  • Provides protection against high-speed particles (120m/s)
  • Clear polycarbonate faceshield
  • High impact resistance
  • Can be used with the H8 Headgear easy faceshield replacement system
TRUSTED_SUSTAINABILITY
Specifications
Product Type Faceshield
Size 230 x 370 mm
Material Polycarbonate