Learn More
Pinnacle Brush™ Wooden Beaker Brushes
Product Code. 12891924
Change view
Click to view available options
Size:
80 x 50 x 150 mm
Brush Circumference: 18 cm
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12891924 Brush Circumference: 18 cm Pack of 6
12801934 80 x 50 x 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12891924 Supplier Pinnacle Brush™ Supplier No. M2117
  • ONLINE EXCLUSIVE PRICE
    23.00 EUR
    20.02 EUR / Pack of 6
    Save 2.98 EUR
    12% Off
Estimated Shipment: 21-12-2026
Features center set bristles. Pinnacle Brush™ Tube Wooden Beaker Brushes offer gentle cleaning for beakers.
  • Rounded shape
  • Wood handle
Specifications
Product Type Beaker Brush
Bristle Material Fiber
Handle Material Wood
Diameter (Metric) 5.7 cm
For Use With (Equipment) Beaker
Length (Metric) 41 cm
Length (Metric) Brush 15.5 cm
Length (Metric) Handle 25.5 cm
Quantity 6 Pk.
Size Brush Circumference: 18 cm