Learn More
Heinz Herenz Wooden Applicator Sterile Single Cotton Bud
Product Code. 30158377
Change view
Click to view available options
Tip Type:
Single End
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Type Unit Size
30158377 Single End Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 30158377 Supplier Heinz Herenz Supplier No. 1030619
Estimated Shipment: 18-12-2026
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Sterile wooden applicator with single cotton bud. Double packed.
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
Specifications
Autoclavable Not Autoclavable
Sterility Sterile
Handle Material Wood
Product Type Cotton Swab
Tip Material Cotton
Tip Type Single End