Learn More
BRAND™ Winkler Pattern Oxygen Flasks
Product Code. 15547841 Shop All Brand Products
Change view
Click to view available options
Volume (Metric) Sample:
250 to 300 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sample Unit Size
15547841 250 to 300 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15547841 Supplier BRAND™ Supplier No. 386048
  • ONLINE EXCLUSIVE PRICE
    67.00 EUR
    58.31 EUR / Pack of 2
    Save 8.69 EUR
    12% Off
View all promotions
Estimated Shipment: 07-12-2026
For the determination of oxygen dissolved in water
  • Soda-lime glass
  • The measured capacity is specified to ± 0,01 ml
  • White labeling area
  • Solid, obliquely cut standard-ground glass stopper can be secured with a fastening clip
  • Each flask is calibrated with its own stopper, hence, stoppers and flasks are not interchangeable
  • Each flask and its stopper is marked with a unique matching identification number
  • TRUSTED_SUSTAINABILITY
    Specifications
    Type NS 19/26
    Includes Glass stopper
    Material Soda-Lime Glass
    Volume (Metric) Sample 250 to 300 mL