Learn More
TERUMO Winged Infusion Sets SURFLO™
Artikelnummer. 16417440
Ansicht ändern
Click to view available options
Needle Gauge:
19 G
23 G
25 G
Packungsgröße:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Needle Gauge Packungsgröße
16417440 25 G Pack of 50
16497430 19 G Pack of 50
16407440 23 G Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16417440 Lieferant TERUMO Lieferanten-Nr. 8SV25NL30
  • ONLINE EXCLUSIVE PRICE
    71.56 EUR
    16.02 EUR / Pack of 50
    Save 55.54 EUR
    77% Off
Alle Werbeaktionen anzeigen
Estimated Shipment: 30-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Designed to ensure stability and grip during venipuncture
  • The Surflo™ Winged Infusion set is intended to introduce or withdraw fluids from the vascular system after vein puncture (18 - 27 G)
  • Highly transparent tubing and adapter for easy recognition of air bubbles and particles
  • The wings are large and flexible for a good grip and secure fixation
  • Reduced pressure on the blood vessel wall due to slightly angled wings
  • Suitable for all applications with a wide product range
TRUSTED_SUSTAINABILITY
Spezifikation
Product Type Infusion Set
Color Orange
Material PVC Tubing, Siliconized Needle
Needle Gauge 25 G
Sterility Sterile
Latex Free Yes