Learn More
Thermo Scientific™ Windshielded Rotor for Cryofuge™ 6000i/8500i
Product Code. 11816460 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
6 x 1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
11816460 6 x 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11816460 Supplier Thermo Scientific™ Supplier No. 75006606
  • ONLINE EXCLUSIVE PRICE
    15029.03 EUR
    11612.00 EUR / Pack of 1
    Save 3417.03 EUR
    22% Off
 Restricted Item
Estimated Shipment: 10-02-2027
Increase lab productivity with the Windshielded Rotor for Cryofuge 6000i/8500i, which comes with a lid and without buckets.
Increase lab productivity with the Thermo Scientific™ Windshielded Rotor for Cryofuge™ 6000i/8500i, which comes with a lid and without buckets.
TRUSTED_SUSTAINABILITY
Specifications
Description Windshielded Swinging Bucket Rotor
Material Stainless Steel Rotor Body, Aluminum Bucket
Type Swinging Bucket Rotor
Capacity 6 x 1000 mL
Max. RCF 6,010 x g (Round), 5,312 x g (Double Blood Bag), 4,984 x g (Double Rectangular)
Max. Speed 4,020 rpm (5,050 rpm with Cryofuge 6000i, Cryofuge 8500i)
K Factor 10,279
For Use With (Equipment) Thermo Scientific Cryofuge 6000i and 8500i Large Capacity Centrifuges
Angle 90°