Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 4 mm Varian Spinner Turbine
Product Code. 15378628
Change view
Click to view available options
For Use With (Equipment):
Agilent/Varian NMR Spectrometer
Model:
V-PEEK-4-NS
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Model Unit Size
15378628 Agilent/Varian NMR Spectrometer V-PEEK-4-NS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15378628 Supplier Bel-Art™ SP Scienceware™ Supplier No. 660000059
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 4 mm Varian Spinner Turbine is manufactured to precision standards that exceed spectrometer products in terms of dimension tolerance, reliability and life span.
  • Cost effective compared to the full VT version for sample changers
  • Allows a portion of the VT air to pass straight past the sample inside the spinner turbine therefore reducing VT gradients and micro sonic flutter due to a high VT flow
  • Includes high temperature external O-rings at the top and bottom for long life and a firm grip on the sample at top and bottom.
  • Can also be used for spinning experiments.
Specifications
For Use With (Equipment) Agilent/Varian NMR Spectrometer
Includes PEEK exchangeable fingers
Model V-PEEK-4-NS