Learn More
Cole-Parmer™ Wildco™ Kemmerer Water Sampler
Product Code. 11784499 Shop All Cole Parmer Products
Change view
Click to view available options
Capacity (Metric):
400 mL
1200 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11784499 1200 mL Each
11774499 400 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11784499 Supplier Cole-Parmer™ Supplier No. 0548510
  • ONLINE EXCLUSIVE PRICE
    1175.00 EUR
    1062.08 EUR / Pack of 1
    Save 112.92 EUR
    9% Off
Estimated Shipment: 15-02-2027
Sample at specific depths between 3 and 600 feet.
  • WildcoKemmerer Water Sampler
  • The messenger activates the unique trip heads that ensure closure in fast flowing streams or turbulent waters, regardless of line angle
  • All models include a plastic carrying case; order messenger and line separately.
  • The 304 SS models have urethane end seals (do not use when mercury concentrations exceed 1 ppm).
Specifications
Product Type Kemmerer Water Sampler
Material Stainless Steel
Description 3 to 600 ft depth
Capacity (Metric) 1200 mL