Learn More
BRAND™ Wide Neck LDPE Bottle
Product Code. 15620479 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15620479 250 mL Each
15600479 50 mL Each
15610479 100 mL Each
15630479 500 mL Each
15640479 1000 mL Each
15650479 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15620479 Supplier BRAND™ Supplier No. 129748
Add to basket
Add to basket
Amber LDPE bottle with wide neck.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Color Amber
Material LDPE
Thread Size (Metric) GL 40
Format Wide Neck
Height (Metric) 126 mm
Diameter (Metric) 62 mm