Learn More
BRAND™ Wide Neck Glass Erlenmeyer Flasks
Product Code. 10089860 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
300 mL
500 mL
1000 mL
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10089860 250 mL Pack of 10
10141681 50 mL Pack of 10
10151871 500 mL Pack of 10
10293391 100 mL Pack of 10
10419912 300 mL Pack of 10
10694352 1000 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10089860 Supplier BRAND™ Supplier No. 92836
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
With beaded rim and graduations
  • Manufactured from Boro 3.3 glass
  • Wide neck for ease of filling
  • Acc. to DIN EN ISO 24450
  • Provided in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer Neck 50 mm
Capacity (Metric) 250 mL
Material Borosilicate Glass (3.3)
Height (Metric) 140 mm
No. per Pack 10
Diameter (Metric) Outer 85 mm