Learn More
Azlon™ Wide Mouth Vented Wash Bottles
Product Code. 15357047
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Closure Color:
Blue
Green
Orange
Red
White
Yellow
Unit Size:
Pack of 5
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Unit Size
15357047 500 mL Blue Pack of 5
15367047 500 mL Orange Pack of 5
15317047 500 mL White Pack of 5
15337047 500 mL Yellow Pack of 5
15347047 500 mL Green Pack of 5
15327047 500 mL Red Pack of 5
15327057 1000 mL Orange Pack of 5
15387047 1000 mL Yellow Pack of 5
15317057 1000 mL Green Pack of 5
15307057 1000 mL Blue Pack of 5
15377047 1000 mL White Pack of 5
15397047 1000 mL Red Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15357047 Lieferant Azlon Lieferanten-Nr. WGW235PVENT
  • ONLINE EXCLUSIVE PRICE
    33.80 EUR
    27.92 EUR / Pack of 5
    Save 5.88 EUR
    17% Off
Alle Werbeaktionen anzeigen
Estimated Shipment: 08-01-2027
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Manufactured from LPDE for strength and flexibilty. Azlon™ Wide Mouth Vented Wash Bottles feature color-coded lids for easy identification.
Manufactured from LPDE for strength and flexibilty. Azlon™ Wide Mouth Vented Wash Bottles feature color coded lids for easy identification.
  • Square shoulder wash bottles
  • Wide neck for easy spill-free filing; patented DripLok™  vapor venting valve prevents solvent drips
  • Color-coded one-piece polypropylene closures maintain shape in use
  • Precision jet-tip aperture for fine control
  • Translucent bottles without print
Spezifikation
Capacity (Metric) 500 mL
Material LDPE
Closure Color Blue
Mouth Wide
Closure Material Polypropylene
Closure Type Screw Cap with Curved Aperture