Learn More
DWK Life Sciences WHEATON™ Vacule™ Lyophilization Ampules
Product Code. 13431429 Shop All Wheaton Science Products Products
Change view
Click to view available options
Diameter (Metric) Outer:
1.2 cm
Unit Size:
Pack of 567
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
13431429 1.2 cm Pack of 567
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13431429 Supplier DWK Life Sciences Supplier No. W651446
  • ONLINE EXCLUSIVE PRICE
    570.00 EUR
    524.81 EUR / Pack of 567
    Save 45.19 EUR
    7% Off
Estimated Shipment: 19-11-2026
WHEATON™ Vacule™ Lyophilization Ampules are designed for small volume samples requiring reliable lyophilization. Made from Type I borosilicate glass, these ampules are available with heat or stopper seal for secure, long-term sample storage.
  • Made from Type I borosilicate glass for superior chemical resistance and thermal stability
  • Designed for lyophilization of small volume samples
  • Available with heat seal or stopper seal closure options
  • Vacule design supports reliable freeze-drying applications
Specifications
Capacity (Metric) 2 mL
Material Borosilicate Glass (Type I)
Product Type Ampule
Color Clear
Diameter (Metric) Outer 1.2 cm
Diameter (English) Outer 0.472 in.
Height (Metric) 86 mm
Height (English) 3.3 in.
Diameter (English)Top 0.3 in.