Learn More
DWK Life Sciences WHEATON™ Straight Plug Rubber Stoppers
Product Code. 10403085 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Gray
Red
Diameter (Metric) Outer:
13 mm
20 mm
Diameter (Metric) Top:
13 mm
20 mm
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Diameter (Metric) Outer Diameter (Metric) Top Unit Size
10403085 Gray 20 mm 20 mm Pack of 1000
11734199 Red 13 mm 13 mm Pack of 1000
12432631 Red 20 mm - Pack of 1000
12462631 Gray 20 mm 20 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10403085 Supplier DWK Life Sciences Supplier No. 224100175
  • ONLINE EXCLUSIVE PRICE
    1480.00 EUR
    1204.36 EUR / Pack of 1000
    Save 275.64 EUR
    18% Off
Estimated Shipment: 21-12-2026
WHEATON™ Straight Plug Rubber Stoppers provide maximum sealing for vials with straight-wall glass finishes. Manufactured from the highest quality raw materials in a variety of sizes and rubber formulations, these autoclavable stoppers deliver exceptional dimensional stability.
  • Straight plug stoppers provide maximum sealing for vials with straight-wall glass finishes
  • Manufactured from the highest quality raw materials
  • Available in a variety of styles, sizes, and rubber formulations
  • Components manufactured to yield exceptional dimensional stability
  • Autoclavable
Specifications
For Use With (Equipment) Vacule Ampules
Autoclavable Autoclavable
Color Gray
Diameter (Metric) Bottom 13 mm
Diameter (Metric) Outer 20 mm
Diameter (Metric) Top 20 mm
Material FEP-coated Chlorobutyl Isoprene Blend
Product Type Straight Plug Stopper
Shape Cylindrical
Sterility Non-sterile
Stopper Number 20