Learn More
DWK Life Sciences Wheaton™ Soxhlet Extraction Apparatus With Standard Taper Ground Joints
Product Code. 12684823 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
Thread Size:
34/45
45/50
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Thread Size Unit Size
12684823 250 mL 45/50 Each
12674823 125 mL 34/45 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12684823 Supplier DWK Life Sciences Supplier No. 416005
  • ONLINE EXCLUSIVE PRICE
    685.00 EUR
    546.21 EUR / Pack of 1
    Save 138.79 EUR
    20% Off
Estimated Shipment: 31-03-2027
For continuous extraction of analytes from a solid into an organic solvent
  • Manufactured from low extractable borosilicate glass
Specifications
Product Type Soxhlet Extraction Apparatus
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Material Borosilicate Glass
Includes Standard taper ground joints, 15mL graduated funnel, anodized aluminum clamp, No. 5 silicone stopper
For Use With (Application) Used for continuous extraction of analytes from a solid into an organic solvent
Certifications/Compliance ASTM E 438
Thread Size 45/50
Standard Taper Joint 24/40