Learn More
DWK Life Sciences Wheaton™ Soxhlet Extraction Apparatus With Standard Taper Ground Joints
Product Code. 12674823 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
Thread Size:
34/45
45/50
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Thread Size unitSize
12674823 125 mL 34/45 Each
12684823 250 mL 45/50 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12674823 Supplier DWK Life Sciences Supplier No. 416000
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
For continuous extraction of analytes from a solid into an organic solvent
  • Manufactured from low extractable borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Product Type Soxhlet Extraction Apparatus
Capacity (Metric) 125 mL
Capacity (English) 4.22 oz.
Material Borosilicate Glass
Includes Standard taper ground joints, 15mL graduated funnel, anodized aluminum clamp, No. 5 silicone stopper
For Use With (Application) Used for continuous extraction of analytes from a solid into an organic solvent
Certifications/Compliance ASTM E 438
Thread Size 34/45
Standard Taper Joint 24/40