Learn More
DWK Life Sciences Wheaton™ Evaporative Flask
Product Code. 12488932 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12488932 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12488932 Supplier DWK Life Sciences Supplier No. 297825
  • ONLINE EXCLUSIVE PRICE
    145.00 EUR
    115.77 EUR / Pack of 1
    Save 29.23 EUR
    20% Off
Estimated Shipment: 31-03-2027
For concentration of trace amounts of analyte in organic solvent
  • 24/40 top and 19/22 bottom clear seal joints
Specifications
Product Type Evaporative Flask
Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Includes 24/40 top and 19/22 bottom clearseal joints
For Use With (Application) Concentration of trace amounts of analyte in organic solvent
Certifications/Compliance ASTM E 438