Learn More
DWK LIFE SCIENCES™ WHEATON™ Three-ball Snyder Column
Product Code. 12431561 Shop All DWK Life Sciences Products
Change view
Click to view available options
Certifications/Compliance:
ASTM E 438
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Certifications/Compliance Unit Size
12431561 ASTM E 438 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12431561 Supplier DWK LIFE SCIENCES™ Supplier No. 384353
  • ONLINE EXCLUSIVE PRICE
    191.00 EUR
    152.34 EUR / Pack of 1
    Save 38.66 EUR
    20% Off
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
WHEATON™ Three-ball Snyder Column is designed for the concentration of trace amounts of analyte in organic solvent, featuring a 24/40 Clear-Seal joint and manufactured from borosilicate glass.
  • Designed for concentration of trace amounts of analyte in organic solvent
  • 24/40 Clear-Seal joint for secure, reliable connections
  • Manufactured from borosilicate glass
  • Three-ball design aids solvent reflux and analyte concentration efficiency
Specifications
For Use With (Equipment) Kuderna-Danish Apparatus
Product Type Three-ball Snyder Column
Includes 24/40 clear seal joint
For Use With (Application) Concentration of trace amounts of analyte in organic solvent
Certifications/Compliance ASTM E 438