Learn More
DWK Life Sciences DURAN™ PURE Bottle, with DIN Thread
BIOPHARMA
Product Code. 15380594 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10 L
10,000 mL
100 mL
1000 mL
20 L
2000 mL
25 mL
250 mL
500 mL
5000 mL
Thread Size (Metric):
GL 25
GL 45
Unit Size:
Each
Pack of 10
Pack of 12
Pack of 120
Pack of 210
Pack of 280
Pack of 30
Pack of 360
Pack of 48
Pack of 540
Pack of 940
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Thread Size (Metric) Unit Size
15380594 100 mL GL 45 Pack of 10
15360594 25 mL GL 25 Pack of 10
15416474 25 mL GL 25 Pack of 940
15436474 100 mL GL 45 Pack of 540
15446474 250 mL GL 45 Pack of 280
15390594 250 mL GL 45 Pack of 10
15336788 500 mL GL 45 Pack of 360
13524960 500 mL GL 45 Pack of 10
13534960 1000 mL GL 45 Pack of 10
15346788 1000 mL - Pack of 210
15356788 2000 mL GL 45 Pack of 120
13544960 2000 mL GL 45 Pack of 10
11945048 5000 mL GL 45 Each
15366788 5000 mL - Pack of 48
11955048 10,000 mL GL 45 Each
15376788 10 L - Pack of 30
15386788 20 L - Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 15380594 Supplier DWK Life Sciences Supplier No. 818012406
  • ONLINE EXCLUSIVE PRICE
    114.45 EUR
    101.78 EUR / Pack of 10
    Save 12.67 EUR
    11% Off
View all promotions
Estimated Shipment: 04-11-2026
DURAN™ PURE Bottle, with DIN Thread is a high-quality Type I borosilicate glass bottle developed for pharmaceutical and biotech industries. Supplied with protective cover, Certificate of Analysis, and full traceability documentation to meet international pharmacopoeia requirements.
  • Made from high-quality Type I borosilicate glass (GL 25/GL 45), available from 25 mL to 20 L
  • Protective cover prevents contamination inside the bottle during storage and transport
  • Single batch delivery for full traceability
  • Certificate of Analysis (CoA) provided with each delivery
  • 100% visual inspection performed on every unit
  • Analysis of hydrolytic resistance according to EP, USP and JP for each batch
  • Change control management with customer notification in case of changes
  • 100% traceability of all manufacturing steps
  • Drug Master File available
  • Meets requirements of international pharmacopoeias
Specifications
Capacity (Metric) 100 mL
Color Clear
For Use With (Application) Manufacturing of pharmaceutical drugs and biotech products; Storage and transport of APIs, bulk drug intermediates, ingredients and media
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 45
Closure Included No
Graduated Yes
Mouth Narrow
Diameter (Metric) 56 mm
Autoclavable Autoclavable
Capacity (English) 3.4 oz.
Shape Round
Disposable Reusable
Product Type Screw Neck Bottle