Learn More
DWK Life Sciences Wheaton™ Diazomethane Generators for Gas Chromatography
Product Code. 12418922 Shop All DWK Life Sciences Products
Change view
Click to view available options
Height (Metric):
19 cm
20 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12418922 20 cm Each
12408922 19 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12418922 Supplier DWK Life Sciences Supplier No. 281155
  • ONLINE EXCLUSIVE PRICE
    214.00 EUR
    166.50 EUR / Pack of 1
    Save 47.50 EUR
    22% Off
Estimated Shipment: 21-04-2027
For generating diazomethane and other gases for gas chromatography, mass sepctroscopy and NMR studies
  • Utilizes clear-seal grease-free joints
  • «O« ring not required
  • Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASMT™ E 438 Type I, Class A requirements
Specifications
Certifications/Compliance USP Type I, ASTM E 438 Type I, Class A
Height (English) 7.8 in.
Height (Metric) 20 cm
Includes O-ring clamp, a 13-425 screw cap with a cut-out top and a ptfe faced rubber liner
Type Diazomethane Generator