Learn More
DWK Life Sciences Wheaton™ Allihn Condensers For Soxhlet Extraction Apparatus
Shop All DWK Life Sciences ProductsDescription
- Wetted parts are manufactured from WHEATON 33 low extractable borosilicate glass
- The flask containing the solvent is heated
- Vapors rise in the larger outside tube
- Vapors enter the water–cooled condenser and liquate
- When the liquid level in the extractor reaches the top of the bent tube, siphoning action returns the extract-enriched solvent to the flask
Compliance
WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Specifications
| Certifications/Compliance | USP Type I and ASTM E438 Type I, Class A |
| For Use With (Application) | Used for continuous extraction of analytes from a solid into an organic solvent |
| For Use With (Equipment) | Soxhlet extraction apparatus |
| Includes | Larger outside tube and bent tube |
| Standard Taper Joint No | 55/50 |
| Material | Borosilicate Glass |
| Type | Allihn Condenser |