Learn More
DWK Life Sciences Wheaton™ Allihn Condensers For Soxhlet Extraction Apparatus
Product Code. 12458942 Shop All Wheaton Science Products Products
Change view
Click to view available options
Standard Taper Joint No:
34/45
55/50
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No unitSize
12458942 34/45 Each
12478942 55/50 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12458942 Supplier DWK Life Sciences Supplier No. 334003
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
Used for continuous extraction of analytes from a solid into an organic solvent
  • Wetted parts are manufactured from WHEATON 33 low extractable borosilicate glass
  • How it works:
    • The flask containing the solvent is heated
    • Vapors rise in the larger outside tube
    • Vapors enter the water–cooled condenser and liquate
    • When the liquid level in the extractor reaches the top of the bent tube, siphoning action returns the extract-enriched solvent to the flask
    Compliance
    WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
    TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
    For Use With (Application) Used for continuous extraction of analytes from a solid into an organic solvent
    For Use With (Equipment) Soxhlet extraction apparatus
    Includes Larger outside tube and bent tube
    Standard Taper Joint No 34/45
    Material Borosilicate Glass
    Type Allihn Condenser