Learn More
Cytiva™ Whatman™ Polyester Drain Discs
Product Code. 11919327
Change view
Click to view available options
Diameter (Metric):
10 mm
22 mm
37 mm
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11919327 22 mm Pack of 100
11909327 10 mm Pack of 100
11929327 37 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11919327 Supplier Cytiva™ Supplier No. 230500
  • ONLINE EXCLUSIVE PRICE
    79.20 EUR
    73.92 EUR / Pack of 100
    Save 5.28 EUR
    6% Off
In Stock
Provides a flat surface to eliminate filter tearing or rupturing. Whatman™ Polyester Drain Discs are used to support membrane filters. They are available in a variety of diameters for use in a range of devices.
For use with membrane hardware where extra support is needed for improved flow rate and throughput. Can be used as a separator between membranes in serial stack filtration applications.
  • Chemically inert polyester
  • 100μm Thickness
  • Binder-free
  • Available in several diameters
TRUSTED_SUSTAINABILITY
Specifications
Material Polyester
For Use With (Application) Membrane filtration, stack filtration
Thickness (Metric) 100 μm
Diameter (Metric) 22 mm