Learn More
Cytiva Whatman™ pH Indicators
Description
Provides rapid determination of pH values. Whatman™ pH Indicators are impregnated with a mixture of several indicators that change color when they come in contact with a particular sample solution.
Determine pH using the supplied color comparison table. Test strips assume a particular color on contact with the sample solution.
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Inexpensive
- Convenient and portable for field use. These pH indicators have been impregnated with a mixture of several indicators
Specifications
Specifications
| pH Range | 1 to 11 |
| Format | Roll |
| Includes | Color comparison chart |
| Quantity | 1 Roll |
| Type | Indicator Paper |