Learn More
Cytiva Whatman™ pH Indicators
Product Code. 11354974
Change view
Click to view available options
pH Range:
0 to 14
1 to 11
2 to 9
Quantity:
1 Roll
100 Pack
200 Pack
Unit Size:
Each
Pack of 100
Pack of 200
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Quantity unitSize
11354974 0 to 14 100 Pack Pack of 100
11364974 2 to 9 100 Pack Pack of 100
11919347 1 to 11 1 Roll Each
11375254 0 to 14 200 Pack Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 11354974 Supplier Cytiva Supplier No. 10362000
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Provides rapid determination of pH values. Whatman™ pH Indicators are impregnated with a mixture of several indicators that change color when they come in contact with a particular sample solution.
Determine pH using the supplied color comparison table. Test strips assume a particular color on contact with the sample solution.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use. These pH indicators have been impregnated with a mixture of several indicators
TRUSTED_SUSTAINABILITY
Specifications
pH Range 0 to 14
Format Strip
Includes Color comparison chart
Quantity 100 Pack
Type Indicator Paper