Learn More
Cytiva Whatman™ Extraction Thimble for DIONEX ASE 200
Product Code. 11979237
Change view
Click to view available options
Wall Thickness:
1 mm
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wall Thickness unitSize
11979237 1 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11979237 Supplier Cytiva Supplier No. 10350106
Excellent mechanical strength and retention
  • Produced from high-quality, alpha cellulose cotton linter
  • Reliable and reproducible analytical results
TRUSTED_SUSTAINABILITY
Specifications
Description Extraction thimbles for DIONEX ASE 200 extraction cell
Capacity (English) 0.37 oz.
Capacity (Metric) 11 mL
Material Cellulose
Wall Thickness 1 mm
For Use With (Application) For excellent mechanical strength and retention
Grade 603
For Use With (Equipment) For dionex ase 200 extraction cell
Max. Temperature (English) 248°F
Max. Temperature (Metric) 130°C
Product Type Extraction Thimble