Learn More
Cytiva Whatman™ 1PS Disposable Phase Separating Paper
Product Code. 11320664
Change view
Click to view available options
Diameter (Metric) Inside:
70 mm
90 mm
110 mm
125 mm
150 mm
185 mm
240 mm
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Unit Size
11320664 90 mm Pack of 100
11330664 110 mm Pack of 100
11340664 125 mm Pack of 100
11318854 240 mm Pack of 100
11360664 185 mm Pack of 100
10272081 150 mm Pack of 100
11310664 70 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11320664 Supplier Cytiva Supplier No. 2200090
  • ONLINE EXCLUSIVE PRICE
    56.50 EUR
    52.21 EUR / Pack of 100
    Save 4.29 EUR
    7% Off
In Stock
Creates a hydrophobic filter that can replace separatory funnel in some applications. Whatman™ 1PS Disposable Phase Separating Paper is impregnated with stable silicone to allow solvent phases to pass through while retaining aqueous phases.
Passes organic solvent phases lighter or heavier than water but retains water phase.
  • Hydrophobic
  • Use folded in conical funnel
  • Disposable
  • Eliminates need for washing and drying separatory funnels, cross-contamination
Order Info
In 7 to 24cm diameter circles.
TRUSTED_SUSTAINABILITY
Specifications
Material Fiberglass/Silicone
For Use With (Application) Liquid phase separation
Type Separator Paper
Volume (English) 0.50 oz.
Volume (Metric) 15 mL
Diameter (English) Inside 3.54 in.
Diameter (Metric) Inside 90 mm