missing translation for 'onlineSavingsMsg'
Learn More

West Nile Virus Envelope Antibody, DyLight 594, Novus Biologicals™

Product Code. 18731613 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18731613 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18731613 Supplier Novus Biologicals Supplier No. NBP241056DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

West Nile Virus Envelope Polyclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen West Nile Virus Envelope
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias DDB1- and CUL4-associated factor 12, CT102, DDB1 and CUL4 associated factor 12, KIAA1892, TCC52, WDR40A
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 16 amino acids at the N terminus of the West Nile virus envelope protein. The immunogen is located within amino acids 350 - 400 of West Nile Virus Envelope Protein. Amino Acid Squence: CPTMGEAHNEKRADPA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.