Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Adapters
Product Code. 10412163 Shop All Welch Ilmvac Products
Change view
Click to view available options
Type:
Adapter, 8-Channel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10412163 Adapter, 8-Channel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10412163 Supplier Welch™ Supplier No. 82885203
Estimated Shipment: 21-12-2026
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
TRUSTED_SUSTAINABILITY
Specifications
For Use With Welch Compact Aspiration/Fitlration Vacuum Station
Material Stainless Steel
No. of Channels 8
Type Adapter, 8-Channel