Learn More
Sartorius™ Weighing Tables
Product Code. 11353284 Shop All Sartorius Products
Change view
Click to view available options
Dimensions (L x W x H):
750 x 900 x 750 mm
900 x 600 x 760 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11353284 750 x 900 x 750 mm Each
10529341 900 x 600 x 760 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11353284 Supplier Sartorius™ Supplier No. YWT09
  • ONLINE EXCLUSIVE PRICE
    2525.00 EUR
    2430.00 EUR / Pack of 1
    Save 95.00 EUR
    3% Off
Estimated Shipment: 07-12-2026
Prevent damage to the surface of weights with these weighing tables.
  • High density of the table's solid material provides excellent support for balances to meet the highest weighing requirements such as those stipulated for calibration of weights using mass comparators
  • Recommended if cleanliness of the weighing area is a major requirement
TRUSTED_SUSTAINABILITY
Spezifikation
Material Wood with cast stone inset
Dimensions (L x W x H) 750 x 900 x 750 mm
Type Weighing Table