Learn More
Microspec™ Wedge Shaped Spreaders
Product Code. 15712125
Change view
Click to view available options
Quantity:
25 Pouches of 20
7500
Individual Pack
Packaging:
Microseal Pouch
Microseal Pouch, Ecopack
Unit Size:
Pack of 250
Pack of 500
Pack of 7500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15712125 25 Pouches of 20 Microseal Pouch Pack of 500
15672767 Individual Pack Microseal Pouch Pack of 250
15702125 7500 Microseal Pouch Pack of 7500
15722125 7500 Microseal Pouch, Ecopack Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15712125 Supplier Microspec™ Supplier No. D10
  • ONLINE EXCLUSIVE PRICE
    30.00 EUR
    25.88 EUR / Pack of 500
    Save 4.12 EUR
    13% Off
Estimated Shipment: 18-12-2026
Wedge shaped to reduce tendency to flip
  • High Impact Polystyrene construction
  • Full length to avoid contamination
  • Disposable
  • Sterile, aseptically manufactured
  • Reduces tendency to flip
Specifications
Disposable Single-use
Color Blue
Length (English) 7.1 in.
Length (Metric) 180 mm
Packaging Microseal Pouch
Quantity 25 Pouches of 20
Material High-impact Polystyrene
Product Type Spreader
Sterility Aseptically Manufactured