Learn More
WAY 100634, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(2-Methoxyphenyl)-N-2-pyridinyl-1-piperazineethanamine |
| CAS | 155204-28-7 |
| Molecular Formula | C18H24N4O |
| Quantity | 1 g |
| Synonym | WAY 100634, 4-(2-Methoxyphenyl)-N-2-pyridinyl-1-piperazineethanamine |
| InChI Formula | InChI=1S/C18H24N4O/c1-23-17-7-3-2-6-16(17)22-14-12-21(13-15-22)11-10-20-18-8-4-5-9-19-18/h2-9H,10-15H2,1H3,(H,19,20) |
| SMILES | COc1ccccc1N2CCN(CCNc3ccccn3)CC2 |
| IUPAC Name | N-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]pyridin-2-amine |
| Molecular Weight (g/mol) | 312.41 |
| Formula Weight | 312.195 |